accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

NH2(R)N

CAS RN 168466-84-0

IUPAC Name: (3R)-1-benzylpiperidin-3-amine
Synonyms: (3R)-1-benzylpiperidin-3-amine
Molecular Weight (g/mol): 190.29
Molecular Formula: C12H18N2
InChi Key: HARWNWOLWMTQCC-GFCCVEGCSA-N
SMILES: N[C@@H]1CCCN(CC2=CC=CC=C2)C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results