accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 16836-95-6

AgCH3SOHOO

CAS RN 16836-95-6

IUPAC Name: 4-methylbenzene-1-sulfonic acid silver
Synonyms: para-toluene sulfonate silver
Molecular Weight (g/mol): 280.07
Molecular Formula: C7H8AgO3S
InChi Key: BRYKBDMLJJLFAB-UHFFFAOYSA-N
SMILES: [Ag].CC1=CC=C(C=C1)S(O)(=O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results