accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 16828-11-8

H2OH2OH2OH2OH2OH2OH2OH2OAlAlOHSOHOOOHSOHOOOHSOHOO

CAS RN 16828-11-8

IUPAC Name: tris(sulfuric acid) dialuminium octahydrate
Synonyms: tris(sulfuric acid) dialuminium octahydrate
Molecular Weight (g/mol): 492.3
Molecular Formula: Al2H22O20S3
InChi Key: JANHUGNIVASISO-UHFFFAOYSA-N
SMILES: O.O.O.O.O.O.O.O.[Al].[Al].OS(O)(=O)=O.OS(O)(=O)=O.OS(O)(=O)=O
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results