accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

H3CH3CCH3OONH(S)FFFOHO

CAS RN 167993-21-7

IUPAC Name: (2S)-2-{[(tert-butoxy)carbonyl]amino}-3-[2-(trifluoromethyl)phenyl]propanoic acid
Synonyms: (2S)-2-[(tert-butoxycarbonyl)amino]-3-[2-(trifluoromethyl)phenyl]propanoic acid
Molecular Weight (g/mol): 333.31
Molecular Formula: C15H18F3NO4
InChi Key: XKMOOODKNPYTEE-NSHDSACASA-N
SMILES: CC(C)(C)OC(=O)N[C@@H](CC1=CC=CC=C1C(F)(F)F)C(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results