accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 1677674-91-7

NHNSNH2Br

CAS RN 1677674-91-7

IUPAC Name: (3-bromopyridin-2-yl)thiourea
Synonyms: 3-bromopyridin-2-ylthiourea
Molecular Weight (g/mol): 232.1
Molecular Formula: C6H6BrN3S
InChi Key: SUOYECWBRSLLQQ-UHFFFAOYSA-N
SMILES: NC(=S)NC1=C(Br)C=CC=N1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results