accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

NH2OOOHO

CAS RN 1676-73-9

IUPAC Name: 2-amino-5-(benzyloxy)-5-oxopentanoic acid
Synonyms: 2-amino-5-(benzyloxy)-5-oxopentanoic acid
Molecular Weight (g/mol): 237.26
Molecular Formula: C12H15NO4
InChi Key: BGGHCRNCRWQABU-UHFFFAOYNA-N
SMILES: NC(CCC(=O)OCC1=CC=CC=C1)C(O)=O
Molecular Weight (g/mol) 
  • (3)
  • (3)
  • (1)
Quantity 
  • (2)
  • (2)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (1)
  • (3)
  • (3)
  • (1)
  • (2)
  • (2)
  • (2)
  • (1)
  • (3)