accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 16753-62-1

CH3OSiCH3OH3CH2C

CAS RN 16753-62-1

IUPAC Name: ethenyldimethoxymethylsilane
Synonyms: ethenyldimethoxymethylsilane
Molecular Weight (g/mol): 132.23
Molecular Formula: C5H12O2Si
InChi Key: ZLNAFSPCNATQPQ-UHFFFAOYSA-N
SMILES: CO[Si](C)(OC)C=C
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results