accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 167479-78-9

H3N(S)NOO

CAS RN 167479-78-9

IUPAC Name: (2S)-2-azaniumyl-3-(4-cyanophenyl)propanoate
Synonyms: (2S)-2-ammonio-3-(4-cyanophenyl)propanoate
Molecular Weight (g/mol): 190.2
Molecular Formula: C10H10N2O2
InChi Key: KWIPUXXIFQQMKN-VIFPVBQESA-N
SMILES: [NH3+][C@@H](CC1=CC=C(C=C1)C#N)C([O-])=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (1)
  • (1)