accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

H3CNOHO

CAS RN 167479-16-5

IUPAC Name: 1-methyl-1H-indole-7-carboxylic acid
Synonyms: 1-methylindole-7-carboxylic acid
Molecular Weight (g/mol): 175.19
Molecular Formula: C10H9NO2
InChi Key: DWKIPLIDZYRILV-UHFFFAOYSA-N
SMILES: CN1C=CC2=CC=CC(C(O)=O)=C12

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results