accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 16728-01-1

H3COHOO

CAS RN 16728-01-1

IUPAC Name: 1-(4-methoxyphenyl)cyclopropane-1-carboxylic acid
Synonyms: 1-(4-methoxyphenyl)cyclopropane-1-carboxylic acid
Molecular Weight (g/mol): 192.21
Molecular Formula: C11H12O3
InChi Key: WCPFQQHADRJANG-UHFFFAOYSA-N
SMILES: COC1=CC=C(C=C1)C1(CC1)C(O)=O
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results