accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 1671-75-6

H3CO

CAS RN 1671-75-6

IUPAC Name: 1-phenylheptan-1-one
Synonyms: 1-heptanone, 1-phenyl-
Molecular Weight (g/mol): 190.29
Molecular Formula: C13H18O
InChi Key: UXMQORVHJMUQFD-UHFFFAOYSA-N
SMILES: CCCCCCC(=O)C1=CC=CC=C1
Molecular Weight (g/mol) 
  • (3)
  • (4)
Quantity 
  • (2)
  • (2)
  • (1)
  • (2)
Boiling Point 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (3)
  • (3)
  • (4)
  • (2)
  • (2)
  • (1)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)