accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CH3NNBr

CAS RN 16681-59-7

IUPAC Name: 2-bromo-1-methyl-1H-imidazole
Synonyms: 2-bromo-1-methylimidazole
Molecular Weight (g/mol): 161
Molecular Formula: C4H5BrN2
InChi Key: BANOTGHIHYMTDL-UHFFFAOYSA-N
SMILES: CN1C=CN=C1Br
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results