accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 166737-85-5

IUPAC Name: 2-amino-3-[(triphenylmethyl)sulfanyl]propanamide
Synonyms: 2-amino-3-[(triphenylmethyl)sulfanyl]propanamide
Molecular Weight (g/mol): 362.49
Molecular Formula: C22H22N2OS
InChi Key: OHWBGKONMFYEKL-UHFFFAOYNA-N
SMILES: NC(CSC(C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1)C(N)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results