accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 16659-88-4

IUPAC Name: 1-[(3,4-dihydroxyphenyl)methyl]-6,7-dihydroxy-1,2,3,4-tetrahydroisoquinolin-2-ium bromide
Synonyms: 1-[(3,4-dihydroxyphenyl)methyl]-6,7-dihydroxy-1,2,3,4-tetrahydroisoquinolin-2-ium bromide
Molecular Weight (g/mol): 368.23
Molecular Formula: C16H18BrNO4
InChi Key: WAADYLVPNMRUKN-UHFFFAOYNA-N
SMILES: [Br-].OC1=CC=C(CC2[NH2+]CCC3=CC(O)=C(O)C=C23)C=C1O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results