accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

NH2NHNN

CAS RN 16617-46-2

IUPAC Name: 5-amino-1H-pyrazole-4-carbonitrile
Synonyms: 3-amino-2H-pyrazole-4-carbonitrile
Molecular Weight (g/mol): 108.1
Molecular Formula: C4H4N4
InChi Key: FFNKBQRKZRMYCL-UHFFFAOYSA-N
SMILES: NC1=C(C=NN1)C#N
Molecular Weight (g/mol) 
  • (3)
  • (4)
Quantity 
  • (2)
  • (2)
  • (1)
  • (2)
Percent Purity 
  • (1)
  • (5)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (1)
  • (3)
  • (4)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (5)
  • (3)