accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 165904-32-5

BOOH3CCH3H3CH3CSiCH3CH3CH3

CAS RN 165904-32-5

IUPAC Name: trimethyl[3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)propyl]silane
Synonyms: trimethyl[3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)propyl]silane
Molecular Weight (g/mol): 242.24
Molecular Formula: C12H27BO2Si
InChi Key: SVYVMKANVAUFQP-UHFFFAOYSA-N
SMILES: CC1(C)OB(CCC[Si](C)(C)C)OC1(C)C
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results