accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

BOOH3CCH3H3CH3C

CAS RN 165904-22-3

IUPAC Name: 4,4,5,5-tetramethyl-2-(2-phenylethyl)-1,3,2-dioxaborolane
Synonyms: 4,4,5,5-tetramethyl-2-(2-phenylethyl)-1,3,2-dioxaborolane
Molecular Weight (g/mol): 232.13
Molecular Formula: C14H21BO2
InChi Key: LVLQNRWCBBIVHR-UHFFFAOYSA-N
SMILES: CC1(C)OB(CCC2=CC=CC=C2)OC1(C)C
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results