accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 16587-71-6

H3CH3CCH3O

CAS RN 16587-71-6

IUPAC Name: 4-(2-methylbutan-2-yl)cyclohexan-1-one
Synonyms: isopentylcyclohexanone
Molecular Weight (g/mol): 168.28
Molecular Formula: C11H20O
InChi Key: DCSKAMGZSIRJAQ-UHFFFAOYSA-N
SMILES: CCC(C)(C)C1CCC(=O)CC1
Boiling Point 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (1)
  • (1)
  • (1)