accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 165253-31-6

OH2N

CAS RN 165253-31-6

IUPAC Name: 1-(oxolan-3-yl)methanamine
Synonyms: 1-(oxolan-3-yl)methanamine
Molecular Weight (g/mol): 101.15
Molecular Formula: C5H11NO
InChi Key: CINJIXGRSTYIHP-UHFFFAOYNA-N
SMILES: NCC1CCOC1
Boiling Point 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results