accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 1648-99-3

FFFSClOO

CAS RN 1648-99-3

IUPAC Name: 2,2,2-trifluoroethane-1-sulfonyl chloride
Synonyms: 2,2,2-trifluoroethanesulfonyl chloride
Molecular Weight (g/mol): 182.54
Molecular Formula: C2H2ClF3O2S
InChi Key: CXCHEKCRJQRVNG-UHFFFAOYSA-N
SMILES: FC(F)(F)CS(Cl)(=O)=O
Boiling Point 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)