accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

BrNHSOO

CAS RN 16468-97-6

IUPAC Name: N-(4-bromophenyl)benzenesulfonamide
Synonyms: N-(4-bromophenyl)benzenesulfonamide
Molecular Weight (g/mol): 312.18
Molecular Formula: C12H10BrNO2S
InChi Key: HHLFFQAEABRCOJ-UHFFFAOYSA-N
SMILES: BrC1=CC=C(NS(=O)(=O)C2=CC=CC=C2)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results