accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

NNOONOOHOHO

CAS RN 164394-23-4

IUPAC Name: 5-nitro-1,10-phenanthroline-2,9-dicarboxylic acid
Synonyms: 5-nitro-1,10-phenanthroline-2,9-dicarboxylic acid
Molecular Weight (g/mol): 313.23
Molecular Formula: C14H7N3O6
InChi Key: XZQXNUFLUBLEIO-UHFFFAOYSA-N
SMILES: OC(=O)C1=CC=C2C=C(C3=CC=C(N=C3C2=N1)C(O)=O)[N+]([O-])=O
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results