accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 16422-67-6

H2OFRb

CAS RN 16422-67-6

IUPAC Name: rubidium(1+) hydrate fluoride
Synonyms: rubidium(1+) hydrate fluoride
Molecular Weight (g/mol): 122.48
Molecular Formula: FH2ORb
InChi Key: CTSZMVONLYPOKW-UHFFFAOYSA-M
SMILES: O.[F-].[Rb+]

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results