CAS RN 16386-93-9
IUPAC Name:
2,2-dimethylpent-4-enoic acid
Synonyms:
2,2-dimethylpent-4-enoic acid
Molecular Weight (g/mol):
128.17
Molecular Formula:
C7H12O2
InChi Key:
BGUAPYRHJPWVEM-UHFFFAOYSA-N
SMILES:
CC(C)(CC=C)C(O)=O
1
–
2
of
2
results
1
2,2-Dimethyl-4-pentenoic acid, 95%
CAS: 16386-93-9 Molecular Formula: C7H12O2 Molecular Weight (g/mol): 128.17 MDL Number: MFCD00012304 InChI Key: BGUAPYRHJPWVEM-UHFFFAOYSA-N Synonym: 2,2-dimethyl-4-pentenoic acid,4-pentenoic acid, 2,2-dimethyl,2,2-dimethyl-pent-4-enoic acid,2,2-dimethyl-4-pentenoicacid,pubchem12797,acmc-1c5lb,ksc495e3r,2,2-dimethyl4-pentenoic acid,2,2-dimethyl-1-4-pentenoic acid 1g PubChem CID: 140065 IUPAC Name: 2,2-dimethylpent-4-enoic acid SMILES: CC(C)(CC=C)C(O)=O