CAS RN 1637562-52-7
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
2-(((Hydroxy-4-methyloctyl)oxy)carbonyl)cyclohexanecarboxylic Acid (Mixture of Diastereomers), TRC
CAS: 1637562-52-7 Molecular Formula: C17H30O5 Molecular Weight (g/mol): 314.42 Synonym: 1,2-Cyclohexanedicarboxylic Acid, Mono-4-methyl-7-hydroxyoctyl Ester,1-(7-Hydroxy-4-methyloctyl) 1,2-cyclohexanedicarboxylate,Cyclohexane-1,2-dicarboxylate mono-(7-hydroxy-4-methyloctyl) ester,1,2-Cyclohexanedicarboxylic acid, 1-(7-hydroxy-4-methyloctyl) ester IUPAC Name: 2-(7-hydroxy-4-methyl-octoxy)carbonylcyclohexanecarboxylic acid SMILES: CC(O)CCC(C)CCCOC(=O)C1CCCCC1C(=O)O