accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 163517-56-4

BOOCH3

CAS RN 163517-56-4

IUPAC Name: 2-(2-methylphenyl)-1,3,2-dioxaborinane
Synonyms: 2-(2-methylphenyl)-1,3,2-dioxaborinane
Molecular Weight (g/mol): 176.02
Molecular Formula: C10H13BO2
InChi Key: UIDYFYHQHHMEKA-UHFFFAOYSA-N
SMILES: CC1=CC=CC=C1B1OCCCO1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results