CAS RN 16343-08-1
IUPAC Name:
hexylboronic acid
Synonyms:
hexylboronic acid
Molecular Weight (g/mol):
129.99
Molecular Formula:
C6H15BO2
InChi Key:
CXSYDLCMCLCOCA-UHFFFAOYSA-N
SMILES:
CCCCCCB(O)O
1
–
1
of
1
results
1
1-Hexylboronic acid, 97%
CAS: 16343-08-1 Molecular Formula: C6H15BO2 Molecular Weight (g/mol): 129.994 MDL Number: MFCD01074641 InChI Key: CXSYDLCMCLCOCA-UHFFFAOYSA-N Synonym: n-hexylboronic acid,1-hexaneboronic acid,boronic acid, b-hexyl,1-hexylboronic acid,n-hexaneboronic acid,boronic acid, hexyl,b-hexyl-boronic acid,n-hexylboronicacid,1-hexylboronic acid c6-b oh 2 PubChem CID: 351064 IUPAC Name: hexylboronic acid SMILES: B(CCCCCC)(O)O