CAS RN 16339-07-4
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
N-Methyl-N’-nitrosopiperazine, TRC
CAS: 16339-07-4 Molecular Formula: C5 H11 N3 O Molecular Weight (g/mol): 129.16 Synonym: Piperazine, 1-methyl-4-nitroso-,1-Methyl-4-nitrosopiperazine,1-Nitroso-4-methylpiperazine,N-Nitroso-N'-methylpiperazine,NSC 523886 IUPAC Name: 1-methyl-4-nitrosopiperazine SMILES: CN1CCN(CC1)N=O