CAS RN 1631-70-5
CAS RN 1631-70-5
IUPAC Name:
benzyldimethylsilyl
Synonyms:
benzyldimethylsilyl
Molecular Weight (g/mol):
149.29
Molecular Formula:
C9H13Si
InChi Key:
SKDKWDVYZOKBPL-UHFFFAOYSA-N
SMILES:
C[Si](C)CC1=CC=CC=C1