accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 16309-45-8

IUPAC Name: 4-(4-nitrophenoxy)benzoic acid
Synonyms: 4-(4-nitrophenoxy)benzoic acid
Molecular Weight (g/mol): 259.22
Molecular Formula: C13H9NO5
InChi Key: XTVBFFGZCMYUJX-UHFFFAOYSA-N
SMILES: OC(=O)C1=CC=C(OC2=CC=C(C=C2)[N+]([O-])=O)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results