accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

ClNNS

CAS RN 16269-66-2

IUPAC Name: 4-chlorothieno[3,2-d]pyrimidine
Synonyms: 4-chlorothieno[3,2-d]pyrimidine
Molecular Weight (g/mol): 170.61
Molecular Formula: C6H3ClN2S
InChi Key: TWTODSLDHCDLDR-UHFFFAOYSA-N
SMILES: ClC1=C2SC=CC2=NC=N1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results