accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 162368-01-6

H3CONOH3C

CAS RN 162368-01-6

IUPAC Name: 1-(4-acetylpiperidin-1-yl)ethan-1-one
Synonyms: 1-(4-acetylpiperidin-1-yl)ethanone
Molecular Weight (g/mol): 169.22
Molecular Formula: C9H15NO2
InChi Key: VFZXZEJGLKOTBW-UHFFFAOYSA-N
SMILES: CC(=O)C1CCN(CC1)C(C)=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results