accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 16205-46-2

IUPAC Name: pyrazolo[1,5-a]pyridine-3-carboxylic acid
Synonyms: pyrazolo[1,5-a]pyridine-3-carboxylic acid
Molecular Weight (g/mol): 162.15
Molecular Formula: C8H6N2O2
InChi Key: HRSDPDBQVZHCRC-UHFFFAOYSA-N
SMILES: OC(=O)C1=C2C=CC=CN2N=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results