accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 16201-96-0

OONNN(E/Z)

CAS RN 16201-96-0

IUPAC Name: 1-[4-(2-phenyldiazen-1-yl)phenyl]-2,5-dihydro-1H-pyrrole-2,5-dione
Synonyms: 1-[4-(2-phenyldiazen-1-yl)phenyl]pyrrole-2,5-dione
Molecular Weight (g/mol): 277.28
Molecular Formula: C16H11N3O2
InChi Key: DVNPYLMPVFDKGZ-UHFFFAOYSA-N
SMILES: O=C1C=CC(=O)N1C1=CC=C(C=C1)N=NC1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results