accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

HOON

CAS RN 1619-37-0

IUPAC Name: (5-phenyl-1,2-oxazol-3-yl)methanol
Synonyms: (5-phenyl-1,2-oxazol-3-yl)methanol
Molecular Weight (g/mol): 175.19
Molecular Formula: C10H9NO2
InChi Key: BPAFLGGUEBMWRN-UHFFFAOYSA-N
SMILES: OCC1=NOC(=C1)C1=CC=CC=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results