accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

H2NS

CAS RN 161886-96-0

IUPAC Name: 3-(thiophen-3-yl)aniline
Synonyms: 3-(thiophen-3-yl)aniline
Molecular Weight (g/mol): 175.25
Molecular Formula: C10H9NS
InChi Key: SHUODWYRFVFGOA-UHFFFAOYSA-N
SMILES: NC1=CC=CC(=C1)C1=CSC=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results