CAS RN 16165-32-5
CAS RN 16165-32-5
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Tris(ethylenediamine)chromium(III) chloride hemiheptahydrate
CAS: 16165-32-5 MDL Number: MFCD00150160 Synonym: Trichlorotris(ethylenediamine)chromium(III)