accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 16110-09-1

ClClN

CAS RN 16110-09-1

IUPAC Name: 2,5-dichloropyridine
Synonyms: 2,5-dichloropyridine
Molecular Weight (g/mol): 147.99
Molecular Formula: C5H3Cl2N
InChi Key: GCTFDMFLLBCLPF-UHFFFAOYSA-N
SMILES: ClC1=CC=C(Cl)N=C1
Boiling Point 
  • (3)
  • (3)
Quantity 
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (3)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (3)