accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 16068-37-4

CH3OSiSiOH3COH3COH3COH3COH3C

CAS RN 16068-37-4

IUPAC Name: 4,4,7,7-tetraethoxy-3,8-dioxa-4,7-disiladecane
Synonyms: 4,4,7,7-tetraethoxy-3,8-dioxa-4,7-disiladecane
Molecular Weight (g/mol): 354.59
Molecular Formula: C14H34O6Si2
InChi Key: IZRJPHXTEXTLHY-UHFFFAOYSA-N
SMILES: CCO[Si](CC[Si](OCC)(OCC)OCC)(OCC)OCC

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results