accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 16066-09-4

H3CSiOSiH3COSiH3CCH3H3COSiCH3CH3H3C

CAS RN 16066-09-4

IUPAC Name:
Synonyms:
Molecular Weight (g/mol): 280.62
Molecular Formula: C8H24O3Si4
InChi Key: OHSYWAVRSCQMHG-UHFFFAOYSA-N
SMILES: C[Si](O[Si](C)O[Si](C)(C)C)O[Si](C)(C)C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results