accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 1604-02-0

CH3HO

CAS RN 1604-02-0

IUPAC Name: 1-propylcyclopentan-1-ol
Synonyms: 1-propylcyclopentan-1-ol
Molecular Weight (g/mol): 128.22
Molecular Formula: C8H16O
InChi Key: GJEILRJIINEWJO-UHFFFAOYSA-N
SMILES: CCCC1(O)CCCC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results