accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CH3SiH3CNHSiCH3H3C

CAS RN 15933-59-2

IUPAC Name:
Synonyms:
Molecular Weight (g/mol): 131.33
Molecular Formula: C4H13NSi2
InChi Key: GJWAPAVRQYYSTK-UHFFFAOYSA-N
SMILES: C[Si](C)N[Si](C)C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results