accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 159239-33-5

HSPOOHHO

CAS RN 159239-33-5

IUPAC Name: (12-sulfanyldodecyl)phosphonic acid
Synonyms: 12-sulfanyldodecylphosphonic acid
Molecular Weight (g/mol): 282.38
Molecular Formula: C12H27O3PS
InChi Key: PVIUMTORLYKRJT-UHFFFAOYSA-N
SMILES: OP(O)(=O)CCCCCCCCCCCCS

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results