CAS RN 15912-68-2
IUPAC Name:
6-fluoro-2-methyl-1,4-dihydroquinolin-4-one
Synonyms:
6-fluoro-2-methyl-1H-quinolin-4-one
Molecular Weight (g/mol):
177.18
Molecular Formula:
C10H8FNO
InChi Key:
BKXCHVFCJZJATJ-UHFFFAOYSA-N
SMILES:
CC1=CC(=O)C2=CC(F)=CC=C2N1
1
–
1
of
1
results
1
6-Fluoro-4-hydroxy-2-methylquinoline, 97%, Thermo Scientific™
CAS: 15912-68-2 Molecular Formula: C10H8FNO Molecular Weight (g/mol): 177.178 MDL Number: MFCD00041442 InChI Key: BKXCHVFCJZJATJ-UHFFFAOYSA-N Synonym: 6-fluoro-2-methylquinolin-4-ol,6-fluoro-2-methylquinolin-4 1h-one,6-fluoro-4-hydroxy-2-methylquinoline,6-fluoro-2-methyl-4-quinolinol,4-quinolinol, 6-fluoro-2-methyl,6-fluoro-2-methyl-quinolin-4-ol,acmc-1aerw,maybridge1_000260,acmc-209j2u,6-fluoro-4-hydroxy quinilidine PubChem CID: 85184 IUPAC Name: 6-fluoro-2-methyl-1H-quinolin-4-one SMILES: CC1=CC(=O)C2=C(N1)C=CC(=C2)F