accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

ONOHNNNOO

CAS RN 1589-62-4

IUPAC Name: 1-cyclohexylidene-2-(2,4-dinitrophenyl)hydrazine
Synonyms: 1-cyclohexylidene-2-(2,4-dinitrophenyl)hydrazine
Molecular Weight (g/mol): 278.27
Molecular Formula: C12H14N4O4
InChi Key: QLWXZRVOHCYKKK-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=CC=C(NN=C2CCCCC2)C(=C1)[N+]([O-])=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results