accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 1589-60-2

HO

CAS RN 1589-60-2

IUPAC Name: 1-phenylcyclohexan-1-ol
Synonyms: α-hydroxycyclohexylbenzene
Molecular Weight (g/mol): 176.26
Molecular Formula: C12H16O
InChi Key: DTTDXHDYTWQDCS-UHFFFAOYSA-N
SMILES: OC1(CCCCC1)C1=CC=CC=C1
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results