accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 1588-83-6

NH2NOOOHO

CAS RN 1588-83-6

IUPAC Name: 4-amino-3-nitrobenzoic acid
Synonyms: 4-amino-3-nitrobenzoic acid
Molecular Weight (g/mol): 182.14
Molecular Formula: C7H6N2O4
InChi Key: ZZNAYFWAXZJITH-UHFFFAOYSA-N
SMILES: NC1=CC=C(C=C1[N+]([O-])=O)C(O)=O
Molecular Weight (g/mol) 
  • (3)
  • (1)
Percent Purity 
  • (3)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (3)
  • (3)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)