accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

H3CONN

CAS RN 15871-85-9

IUPAC Name: 6-methoxypyridine-3-carbonitrile
Synonyms: 6-methoxypyridine-3-carbonitrile
Molecular Weight (g/mol): 134.14
Molecular Formula: C7H6N2O
InChi Key: DFPYAQAFVHRSAG-UHFFFAOYSA-N
SMILES: COC1=NC=C(C=C1)C#N

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results