accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 15871-57-5

CH3H3CNH2H3CH3CNOOHO

CAS RN 15871-57-5

IUPAC Name: (4-amino-4-carboxy-2,2,6,6-tetramethylpiperidin-1-yl)oxidanyl
Synonyms: TOAC
Molecular Weight (g/mol): 215.27
Molecular Formula: C10H19N2O3
InChi Key: FCTFGDVLBOGMPH-UHFFFAOYSA-N
SMILES: CC1(C)CC(N)(CC(C)(C)N1[O])C(O)=O
Quantity 
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)