accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 15861-36-6

NHN

CAS RN 15861-36-6

IUPAC Name: 1H-indole-6-carbonitrile
Synonyms: indole-6-cyano-
Molecular Weight (g/mol): 142.16
Molecular Formula: C9H6N2
InChi Key: SZSZDBFJCQKTRG-UHFFFAOYSA-N
SMILES: N#CC1=CC=C2C=CNC2=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results